Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0700014615.mm.1 |
TC0700001731.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014615.mm.1 |
NM_025899.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014615.mm.1 |
NM_025899.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014619.mm.1 |
TC0700001731.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014619.mm.1 |
NM_025899.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014619.mm.1 |
NM_025899.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014618.mm.1 |
TC0700001731.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014618.mm.1 |
NM_025899.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014618.mm.1 |
NM_025899.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014613.mm.1 |
TC0700001731.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0700014613.mm.1 |
NM_025899.2.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0700014613.mm.1 |
NM_025899.2 |
Refseq |