Assay Details
| Gene Symbol: Uqcrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025899.2 | NP_080175.1 | 3 - 4 | 369 | 77 | |
| GenBank mRNA | AK003975.1 | - | 3 - 4 | 342 | 77 | |
| AK004492.1 | - | 3 - 4 | 344 | 77 | ||
| AK005151.1 | - | 3 - 4 | 353 | 77 | ||
| AK007669.1 | - | 2 - 3 | 233 | 77 | ||
| AK075856.1 | - | 3 - 4 | 313 | 77 | ||
| AK077583.1 | - | 3 - 4 | 318 | 77 | ||
| AK088103.1 | - | 3 - 4 | 323 | 77 | ||
| AK167377.1 | - | 3 - 4 | 323 | 77 | ||
| BC003423.1 | - | 3 - 4 | 341 | 2609295 | 77 | |
More Information
Updated Mapping Information
Set Membership: |
|

Uqcrc2
Genomic Map