Assay Details
| Gene Symbol: Prr14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145589.2 | NP_663564.2 | 4 - 5 | 842 | 75 | |
| GenBank mRNA | AK133884.1 | - | 4 - 5 | 806 | 75 | |
| AK139559.1 | - | 5 - 6 | 617 | 75 | ||
| AK145816.1 | - | 5 - 6 | 622 | 75 | ||
| AK151203.1 | - | 5 - 6 | 674 | 75 | ||
| BC055033.1 | - | 4 - 5 | 842 | 6310272 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Prr14
Genomic Map