Assay Details
| Gene Symbol: Hoxd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073086.1 | 1 - 2 | 251 | 81 | ||
| GenBank mRNA | AK054090.1 | - | 1 - 2 | 252 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hoxd11| Gene Symbol: Hoxd11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_073086.1 | 1 - 2 | 251 | 81 | ||
| GenBank mRNA | AK054090.1 | - | 1 - 2 | 252 | 81 | |
Set Membership: |
|