| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200010649.mm.1 |
NM_001166024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010649.mm.1 |
NM_001166024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010649.mm.1 |
NM_023854.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010649.mm.1 |
NM_023854.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010649.mm.1 |
TC0200001375.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200010664.mm.1 |
NM_001166024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010664.mm.1 |
NM_001166024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010664.mm.1 |
TC0200001375.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200010654.mm.1 |
NM_001166024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010654.mm.1 |
NM_001166024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010654.mm.1 |
NM_023854.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010654.mm.1 |
NM_023854.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010654.mm.1 |
TC0200001375.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200010660.mm.1 |
NM_001166024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010660.mm.1 |
NM_001166024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010660.mm.1 |
NM_023854.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010660.mm.1 |
NM_023854.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010660.mm.1 |
TC0200001375.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200010662.mm.1 |
NM_001166024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010662.mm.1 |
NM_001166024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010662.mm.1 |
NM_023854.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010662.mm.1 |
NM_023854.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010662.mm.1 |
TC0200001375.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200010653.mm.1 |
NM_001166024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010653.mm.1 |
NM_001166024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010653.mm.1 |
NM_023854.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010653.mm.1 |
NM_023854.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010653.mm.1 |
TC0200001375.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200010658.mm.1 |
NM_001166024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010658.mm.1 |
NM_001166024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010658.mm.1 |
NM_023854.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010658.mm.1 |
NM_023854.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010658.mm.1 |
TC0200001375.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200010656.mm.1 |
NM_001166024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010656.mm.1 |
NM_001166024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010656.mm.1 |
NM_023854.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010656.mm.1 |
NM_023854.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010656.mm.1 |
TC0200001375.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200010661.mm.1 |
NM_001166024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010661.mm.1 |
NM_001166024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010661.mm.1 |
NM_023854.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010661.mm.1 |
NM_023854.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010661.mm.1 |
TC0200001375.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200010651.mm.1 |
NM_001166024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010651.mm.1 |
NM_001166024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010651.mm.1 |
NM_023854.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010651.mm.1 |
NM_023854.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010651.mm.1 |
TC0200001375.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200010659.mm.1 |
NM_001166024.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010659.mm.1 |
NM_001166024.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010659.mm.1 |
NM_023854.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010659.mm.1 |
NM_023854.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200010659.mm.1 |
TC0200001375.mm.1 |
MTA TC |