Assay Details
| Gene Symbol: Dnajc24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033993.1 | 2 - 3 | 609 | 150 | ||
| GenBank mRNA | AK011571.1 | - | 2 - 3 | 323 | 150 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dnajc24
Genomic Map