Assay Details
| Gene Symbol: Kcnj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK046778.1 | - | 1 - 2 | 791 | 95 | |
| BC068167.1 | - | 2 - 3 | 1670 | 30543982 | 95 | |
| BC103671.1 | - | 1 - 2 | 746 | 95 | ||
| BC103673.1 | - | 1 - 2 | 746 | 40046971 | 95 | |
| BC103674.1 | - | 1 - 2 | 746 | 40046969 | 95 | |
| D45022.1 | - | 1 - 2 | 754 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Kcnj3
Genomic Map