Assay Details
| Gene Symbol: Slc22a18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042760.1 | NP_001036225.1 | 1 - 2 | 126 | 71 | |
| GenBank mRNA | AB012084.1 | - | 1 - 2 | 126 | 71 | |
| AK002460.1 | - | 1 - 2 | 67 | 71 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB012084.1, NM_001042760.1 transcripts.
Set Membership: |
|

Slc22a18
Genomic Map