Assay Details
| Gene Symbol: Paox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC033913.1 | - | 2 - 3 | 146 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC033913.1 transcripts.
Set Membership: |
|

Paox
Genomic Map