Assay Details
| Gene Symbol: Phkg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026888.3 | NP_081164.2 | 1 - 2 | 111 | 91 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_026888.3 transcripts.
Set Membership: |
|

Phkg2
Genomic Map