Assay Details
| Gene Symbol: Phkg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026888.3 | NP_081164.2 | 2 - 3 | 221 | 62 | |
| GenBank mRNA | AK005277.1 | - | 2 - 3 | 166 | 62 | |
| AK079680.1 | - | 2 - 3 | 340 | 62 | ||
| AK089062.1 | - | 2 - 3 | 168 | 62 | ||
| BC138913.1 | - | 2 - 3 | 135 | 8861935 | 62 | |
| BC145734.1 | - | 2 - 3 | 135 | 40131037 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phkg2
Genomic Map