Assay Details
| Gene Symbol: Phkg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026888.3 | NP_081164.2 | 4 - 5 | 456 | 94 | |
| GenBank mRNA | AK005277.1 | - | 4 - 5 | 401 | 94 | |
| BC138913.1 | - | 4 - 5 | 370 | 8861935 | 94 | |
| BC145734.1 | - | 4 - 5 | 370 | 40131037 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Phkg2
Genomic Map