Assay Details
| Gene Symbol: Phkg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026888.3 | NP_081164.2 | 1 - 2 | 111 | 73 | |
| GenBank mRNA | AK005277.1 | - | 1 - 2 | 56 | 73 | |
| AK089062.1 | - | 1 - 2 | 58 | 73 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK005277.1, NM_026888.3 transcripts.
Set Membership: |
|

Phkg2
Genomic Map