Assay Details
| Gene Symbol: Nsmce4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162855.1 | NP_001156327.1 | 3 - 4 | 83 | ||
| GenBank mRNA | BC032990.1 | - | 8 - 9 | 834 | 83 | |
| BC051572.1 | - | 8 - 9 | 849 | 83 | ||
| BE951712.1 | - | 3 - 4 | 83 | |||
More Information
Set Membership: |
|

Nsmce4a
Genomic Map