Assay Details
| Gene Symbol: Nsmce4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162855.1 | NP_001156327.1 | 8 - 9 | 1086 | 89 | |
| GenBank mRNA | AK010349.1 | - | 8 - 9 | 1086 | 89 | |
| AK010795.1 | - | 8 - 9 | 1052 | 89 | ||
| AK189457.1 | - | 4 - 5 | 283 | 89 | ||
| BC032990.1 | - | 3 - 4 | 89 | |||
| BC051572.1 | - | 3 - 4 | 89 | |||
| BC089629.1 | - | 5 - 6 | 444 | 89 | ||
More Information
Set Membership: |
|

Nsmce4a
Genomic Map