Assay Details
| Gene Symbol: Nsmce4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162855.1 | NP_001156327.1 | 9 - 10 | 104 | ||
| GenBank mRNA | AK010349.1 | - | 9 - 10 | 104 | ||
| AK010795.1 | - | 9 - 10 | 104 | |||
| AK189457.1 | - | 5 - 6 | 105 | |||
| BC032990.1 | - | 2 - 3 | 260 | 104 | ||
| BC051572.1 | - | 2 - 3 | 275 | 104 | ||
| BC089629.1 | - | 6 - 7 | 104 | |||
More Information
Set Membership: |
|

Nsmce4a
Genomic Map