Assay Details
| Gene Symbol: Pwwp2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033206.2 | NP_001028378.1 | 2 - 3 | 1549 | 60 | |
| GenBank mRNA | AK158245.1 | - | 2 - 3 | 1549 | 60 | |
| AK160932.1 | - | 2 - 3 | 1471 | 60 | ||
| AK207913.1 | - | 1 - 2 | 336 | 60 | ||
| BC116669.1 | - | 2 - 3 | 1549 | 40107062 | 60 | |
| BC116720.1 | - | 2 - 3 | 1545 | 40107061 | 60 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pwwp2b
Genomic Map