Assay Details
| Gene Symbol: Slc5a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177624.1 | NP_001171095.1 | 11 - 12 | 1614 | 81 | |
| GenBank mRNA | AK085356.1 | - | 11 - 12 | 1609 | 81 | |
| AK128951.1 | - | 11 - 12 | 1606 | 81 | ||
| AK144004.1 | - | 11 - 12 | 1547 | 81 | ||
| AK165419.1 | - | 11 - 12 | 1607 | 81 | ||
| AY964639.1 | - | 11 - 12 | 1317 | 81 | ||
| BC121791.1 | - | 11 - 12 | 1310 | 81 | ||
| BC141255.1 | - | 11 - 12 | 1542 | 9055867 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc5a12
Genomic Map