Assay Details
| Gene Symbol: Slc5a12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003915.2 | NP_001003915.1 | 9 - 10 | 1458 | 73 | |
| NM_001177624.1 | NP_001171095.1 | 9 - 10 | 1458 | 73 | ||
| GenBank mRNA | AF529222.1 | - | 9 - 10 | 1466 | 73 | |
| AK085356.1 | - | 9 - 10 | 1452 | 73 | ||
| AK128951.1 | - | 9 - 10 | 1450 | 73 | ||
| AK144004.1 | - | 9 - 10 | 1391 | 73 | ||
| AK165419.1 | - | 9 - 10 | 1451 | 73 | ||
| AY964639.1 | - | 9 - 10 | 1161 | 73 | ||
| BC121791.1 | - | 9 - 10 | 1154 | 73 | ||
| BC141255.1 | - | 9 - 10 | 1386 | 9055867 | 73 | |
More Information
Set Membership: |
|

Slc5a12
Genomic Map