Assay Details
| Gene Symbol: Olfr665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146814.1 | NP_667025.1 | 1 | 841 | 82 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2095683 | NM_146814.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 901385 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P160740 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2095683 | NM_146814.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P216990 | NM_146814.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P160740 | - | - |
| Whole Mouse Genome G4122A | A_51_P216990 | NM_146814.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P134685 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P160740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389764 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389666 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389762 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389702 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389745 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389671 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389747 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389716 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389669 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389740 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389703 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389706 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389701 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389661 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389662 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389741 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389677 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389738 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389733 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389759 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389688 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389720 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389659 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389705 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389690 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389699 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389752 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389736 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389713 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389679 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389724 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P25389683 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389745 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389696 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389685 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389680 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389724 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06280436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389750 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389698 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06355890 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389678 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389736 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06247726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389749 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06477444 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06654893 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389740 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389720 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389694 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389681 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389690 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389756 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389691 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06238577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389706 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389703 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06247725 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389719 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389747 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389693 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389737 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389729 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389741 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06477459 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389679 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389752 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389754 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06477464 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389711 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389676 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389677 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25389733 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10555900 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10555899 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 567978 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 2970162 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002970162 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1223342 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1223342 | - | - |
| mouse-6 v1.1 (Illumina) | scl32249.1.1_271-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2970162 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl32249.1.1_271-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2970162 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1223342 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1223342 | - | - |

Olfr665
Genomic Map