Assay Details
| Gene Symbol: Lnp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110209.1 | NP_001103679.1 | 10 - 11 | 990 | 104 | |
| NM_027133.3 | NP_081409.1 | 10 - 11 | 990 | 104 | ||
| GenBank mRNA | AK014897.1 | - | 10 - 11 | 887 | 104 | |
| AK048501.1 | - | 1 - 2 | 1233 | 104 | ||
| AK082900.1 | - | 9 - 10 | 915 | 104 | ||
| AK136506.1 | - | 10 - 11 | 965 | 104 | ||
| AK137445.1 | - | 10 - 11 | 991 | 104 | ||
| AK173241.1 | - | 10 - 11 | 922 | 104 | ||
| AY312281.1 | - | 10 - 11 | 911 | 104 | ||
| BC057961.1 | - | 10 - 11 | 917 | 5353629 | 104 | |
| BC060153.1 | - | 9 - 10 | 829 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lnp
Genomic Map