| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1700001439.mm.1 |
NR_033586.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001439.mm.1 |
NR_033586.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001439.mm.1 |
TC1700000182.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001443.mm.1 |
NM_024203.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001443.mm.1 |
NM_024203.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001443.mm.1 |
NR_033586.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001443.mm.1 |
NR_033586.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001443.mm.1 |
TC1700000182.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001448.mm.1 |
NM_024203.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001448.mm.1 |
NM_024203.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001448.mm.1 |
NR_033586.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001448.mm.1 |
NR_033586.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001448.mm.1 |
TC1700000182.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1700001449.mm.1 |
NR_033586.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001449.mm.1 |
NR_033586.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1700001449.mm.1 |
TC1700000182.mm.1 |
MTA TC |