Assay Details
| Gene Symbol: Gyltl1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166633.2 | NP_001160105.1 | 3 - 4 | 329 | 59 | |
| NM_172670.3 | NP_766258.2 | 3 - 4 | 329 | 59 | ||
| GenBank mRNA | AK053214.1 | - | 3 - 4 | 341 | 59 | |
| AK081797.1 | - | 3 - 4 | 306 | 59 | ||
| AY742914.1 | - | 3 - 4 | 305 | 59 | ||
| AY742915.1 | - | 3 - 4 | 305 | 59 | ||
| BC096655.1 | - | 2 - 3 | 256 | 5057489 | 59 | |
| HM141800.1 | - | 2 - 3 | 158 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Gyltl1b
Genomic Map