Assay Details
| Gene Symbol: Tjap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028751.3 | NP_083027.1 | 2 - 3 | 163 | 62 | |
| GenBank mRNA | AK002882.2 | - | 2 - 3 | 150 | 62 | |
| AK190015.1 | - | 1 - 2 | 58 | 62 | ||
| BC023316.1 | - | 2 - 3 | 149 | 3993257 | 62 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002882.2, BC023316.1, NM_028751.3 transcripts.
Set Membership: |
|

Tjap1
Genomic Map