Assay Details
| Gene Symbol: Tjap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252474.1 | NP_001239403.1 | 3 - 4 | 254 | 72 | |
| NM_001252475.1 | NP_001239404.1 | 2 - 3 | 160 | 72 | ||
| GenBank mRNA | AK164925.1 | - | 3 - 4 | 239 | 72 | |
| AK190015.1 | - | 3 - 4 | 279 | 72 | ||
| AK211075.1 | - | 2 - 3 | 72 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001252474.1, NM_001252475.1 transcripts.
Set Membership: |
|

Tjap1
Genomic Map