Assay Details
| Gene Symbol: Tubgcp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001290824.1 | NP_001277753.1 | 8 - 9 | 1116 | 127 | |
| NM_153387.3 | NP_700436.1 | 8 - 9 | 1116 | 127 | ||
| NR_077245.2 | 8 - 9 | 1116 | 127 | |||
| NR_110985.1 | 8 - 9 | 1116 | 127 | |||
| GenBank mRNA | AK016563.1 | - | 8 - 9 | 1159 | 127 | |
| AK038921.1 | - | 8 - 9 | 1075 | 127 | ||
| AK045305.1 | - | 8 - 9 | 1009 | 127 | ||
| AK046132.1 | - | 8 - 9 | 1046 | 127 | ||
| AK050250.1 | - | 8 - 9 | 1117 | 127 | ||
| AK051517.1 | - | 8 - 9 | 1017 | 127 | ||
| AK152984.1 | - | 8 - 9 | 1075 | 127 | ||
| AK153171.1 | - | 8 - 9 | 1075 | 127 | ||
| AK197101.1 | - | 2 - 3 | 337 | 127 | ||
| AK217929.1 | - | 2 - 3 | 367 | 127 | ||
| BC029106.1 | - | 8 - 9 | 1080 | 3711922 | 127 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tubgcp4
Genomic Map