Assay Details
| Gene Symbol: Gm35692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK053380.1 | - | 2 - 3 | 335 | 105 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | RIKEN cDNA F630040K05 gene | F630040K05Rik | -19.8kb | Anti-sense | Chr. 17 - 45734506 - 45786357 on NCBI Build GRCm38 | |
| Downstream | RIKEN cDNA 4930542M03 gene | 4930542M03Rik | 72.0kb | Sense | Chr. 17 - 45880119 - 45881452 on NCBI Build GRCm38 |
More Information
Set Membership: |
|

Gm35692
Genomic Map