Assay Details
| Gene Symbol: Gm35725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK039641.1 | - | 1 - 2 | 92 | 84 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | DEAD (Asp-Glu-Ala-Asp) box polypeptide 1 | Ddx1 |
AA409185, DBP-RB |
-364.3kb | Sense | Chr. 12 - 13219307 - 13249174 on NCBI Build GRCm38 |
| Downstream |
family with sequence similarity 84, member A |
Fam84a |
2310003N02Rik, 4731402F03Rik, AW125753, Nse1 |
520.0kb | Sense | Chr. 12 - 14147598 - 14152038 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

Gm35725
Genomic Map