Assay Details
| Gene Symbol: Fsip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027759.3 | NP_082035.1 | 4 - 5 | 636 | 138 | |
| GenBank mRNA | AF448787.1 | - | 4 - 5 | 636 | 138 | |
| AK005924.1 | - | 4 - 5 | 627 | 138 | ||
| AK017026.1 | - | 4 - 5 | 625 | 138 | ||
| BC125024.1 | - | 4 - 5 | 528 | 40056533 | 138 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fsip1
Genomic Map