Assay Details
| Gene Symbol: Nctc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_002452.2 | 1 - 2 | 199 | 83 | ||
| GenBank mRNA | AI594936.1 | - | 1 - 2 | 199 | 83 | |
| AK035596.1 | - | 1 - 2 | 198 | 83 | ||
| AK036720.1 | - | 1 - 2 | 199 | 83 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitochondrial ribosomal protein L23 | Mrpl23 |
L23mrp, Rpl23, Rpl23l |
-14.3kb | Anti-sense | Chr. 7 - 142533030 - 142540747 on NCBI Build GRCm38 |
| Downstream |
H19, imprinted maternally expressed transcript |
H19 | AI747191 | 16.9kb | Sense | Chr. 7 - 142575532 - 142578146 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_66_P106095 | AK036720.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 154844 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 815148 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 906977 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 786437 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 6942 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729511 | AK033856.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729511 | AK049048.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729511 | AK049938.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729511 | AK085603.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729511 | AK141578.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729511 | BC093486.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 729511 | NM_178768.4 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 470941 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 701429 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 861451 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 649530 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 7260 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 83734 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_66_P106095 | AK036720.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P01657693 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P06399594 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P135090 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_67_P06399594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595537 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06576671 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06576665 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595555 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595521 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595509 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06576645 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595570 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595519 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595507 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06029434 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595514 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595615 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595500 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06576620 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06576617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595586 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595574 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595546 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595606 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595559 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06287774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595533 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595577 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06029436 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595552 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595558 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595591 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06576629 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06188367 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595566 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595613 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595598 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595497 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595557 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595531 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595503 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595505 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595542 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06287773 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595593 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595604 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595496 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595553 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06251712 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595587 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595528 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06188383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595618 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595529 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595539 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595616 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595526 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595524 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06010118 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06010124 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595565 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595576 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06233875 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595589 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06576630 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06251701 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595590 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595592 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06029450 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595597 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06287789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595608 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595594 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595611 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06576653 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06233877 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595513 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06576636 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06010119 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595601 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595527 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595548 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595617 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595549 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595612 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06188378 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06576675 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595569 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06029439 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595536 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595581 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595518 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P06010120 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P25595573 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Clariom S Array | PSR0700038496.mm.1 | NR_002452.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700038496.mm.1 | NR_002452.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700038496.mm.1 | TC0700004568.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700038498.mm.1 | NR_002452.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700038498.mm.1 | NR_002452.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700038498.mm.1 | TC0700004568.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0700038499.mm.1 | NR_002452.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700038499.mm.1 | NR_002452.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0700038499.mm.1 | TC0700004568.mm.1 | MTA TC |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 312133 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2523613 | - | - |

Nctc1
Genomic Map