Assay Details
| Gene Symbol: 1700012D14Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015573.2 | 1 - 2 | 296 | 109 | ||
| GenBank mRNA | AK005898.1 | - | 1 - 2 | 298 | 109 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
eukaryotic translation initiation factor 4, gamma 2 |
Eif4g2 |
AA589388, DAP-5, E130105L11Rik, Nat1, Natm1, p97 |
-37.9kb | Sense | Chr. 7 - 111067985 - 111083030 on NCBI Build GRCm38 |
| Downstream | UDP-N-acetyl-alpha-D-galactosamine:polypeptide N-acetylgalactosaminyltransferase 18 | Galnt18 |
2900011G21Rik, BC024988, Galntl4 |
349.0kb | Sense | Chr. 7 - 111471656 - 111780067 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

1700012D14Rik
Genomic Map