Assay Details
| Gene Symbol: Ndst2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010811.2 | NP_034941.2 | 10 - 11 | 2718 | 89 | |
| GenBank mRNA | AF074925.1 | - | 8 - 9 | 1968 | 89 | |
| AK150100.1 | - | 10 - 11 | 2499 | 89 | ||
| BC050829.1 | - | 10 - 11 | 2483 | 89 | ||
| BC110479.1 | - | 8 - 9 | 2054 | 40048114 | 89 | |
| BC110480.1 | - | 8 - 9 | 2116 | 40048115 | 89 | |
| U02304.1 | - | 9 - 10 | 2480 | 89 | ||
| X75885.1 | - | 8 - 9 | 2122 | 89 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ndst2
Genomic Map