Assay Details
| Gene Symbol: 5830432E09Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015548.1 | 1 - 2 | 530 | 77 | ||
| GenBank mRNA | AK017962.1 | - | 1 - 2 | 531 | 77 | |
| AK020025.1 | - | 1 - 2 | 528 | 77 | ||
| AK088640.1 | - | 1 - 2 | 501 | 77 | ||
| AK148045.1 | - | 374 | 77 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA 1962 | Mir1962 |
mir-1962, mmu-mir-1962 |
-84.6kb | Anti-sense | Chr. 7 - 135566162 - 135566282 on NCBI Build GRCm38 |
| Downstream | antigen identified by monoclonal antibody Ki 67 | Mki67 |
D630048A14Rik, Ki-67, Ki67 |
37.9kb | Sense | Chr. 7 - 135689782 - 135716408 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

5830432E09Rik
Genomic Map