Assay Details
| Gene Symbol: Gm14023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040371.1 | 1 - 2 | 34 | 70 | ||
| GenBank mRNA | AK042010.1 | - | 1 - 2 | 35 | 70 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cytoskeleton associated protein 2-like | Ckap2l |
2010016H04Rik, 2610318C08Rik, AV070319, radmis, rdm |
-140.0 | Anti-sense | Chr. 2 - 129268210 - 129297230 on NCBI Build GRCm38 |
| Downstream | interleukin 1 alpha | Il1a | Il-1a | 772.0 | Anti-sense | Chr. 2 - 129299609 - 129310901 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm14023
Genomic Map