Assay Details
| Gene Symbol: Gm14023 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040371.1 | 3 - 4 | 279 | 108 | ||
| GenBank mRNA | AK042010.1 | - | 3 - 4 | 280 | 108 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | interleukin 1 alpha | Il1a | Il-1a | Anti-sense | Chr. 2 - 129299609 - 129310901 on NCBI Build GRCm38 | |
| Downstream | interleukin 1 beta | Il1b |
IL-1beta, Il-1b |
57.5kb | Anti-sense | Chr. 2 - 129363843 - 129371173 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm14023
Genomic Map