| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0200015440.mm.1 |
NR_040371.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015440.mm.1 |
NR_040371.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015440.mm.1 |
TC0200001954.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015441.mm.1 |
NR_040371.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015441.mm.1 |
NR_040371.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015441.mm.1 |
TC0200001954.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015442.mm.1 |
NR_040371.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015442.mm.1 |
NR_040371.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015442.mm.1 |
TC0200001954.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015439.mm.1 |
NR_040371.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015439.mm.1 |
NR_040371.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015439.mm.1 |
TC0200001954.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0200015438.mm.1 |
NR_040371.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015438.mm.1 |
NR_040371.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0200015438.mm.1 |
TC0200001954.mm.1 |
MTA TC |