Assay Details
| Gene Symbol: Mettl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145524.3 | NP_663499.2 | 3 - 4 | 477 | 126 | |
| GenBank mRNA | AK042669.1 | - | 3 - 4 | 455 | 126 | |
| AK087927.1 | - | 3 - 4 | 407 | 126 | ||
| BC004636.1 | - | 3 - 4 | 398 | 3585719 | 126 | |
| BC057960.1 | - | 3 - 4 | 344 | 5354784 | 126 | |
| DQ093357.1 | - | 3 - 4 | 454 | 126 | ||
| DQ093358.1 | - | 3 - 4 | 454 | 126 | ||
| DQ230973.1 | - | 2 - 3 | 245 | 126 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC004636.1, BC057960.1 transcripts.
Set Membership: |
|

Mettl8
Genomic Map