Assay Details
| Gene Symbol: Mettl8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK087927.1 | - | 5 - 6 | 818 | 62 | |
| BC004636.1 | - | 6 - 7 | 859 | 3585719 | 62 | |
| BC057960.1 | - | 6 - 7 | 805 | 5354784 | 62 | |
| DQ093357.1 | - | 5 - 6 | 865 | 62 | ||
| DQ093358.1 | - | 6 - 7 | 915 | 62 | ||
| DQ230973.1 | - | 5 - 6 | 706 | 62 | ||
| DQ230974.1 | - | 5 - 6 | 478 | 62 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mettl8
Genomic Map