Assay Details
| Gene Symbol: Trmt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175113.3 | NP_780322.2 | 5 - 6 | 664 | 96 | |
| GenBank mRNA | AK017672.1 | - | 5 - 6 | 606 | 96 | |
| AK028755.1 | - | 5 - 6 | 664 | 96 | ||
| AK129300.1 | - | 5 - 6 | 631 | 96 | ||
| AK164396.1 | - | 5 - 6 | 597 | 96 | ||
| AK165976.1 | - | 5 - 6 | 616 | 96 | ||
| AK167222.1 | - | 5 - 6 | 630 | 96 | ||
| AK194631.1 | - | 3 - 4 | 293 | 96 | ||
| BC052648.1 | - | 5 - 6 | 582 | 30030801 | 96 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt6
Genomic Map