Assay Details
| Gene Symbol: Trmt6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175113.3 | NP_780322.2 | 7 - 8 | 1147 | 88 | |
| GenBank mRNA | AK014359.1 | - | 7 - 8 | 1263 | 88 | |
| AK028755.1 | - | 7 - 8 | 1147 | 88 | ||
| AK129300.1 | - | 7 - 8 | 1114 | 88 | ||
| AK164396.1 | - | 7 - 8 | 1080 | 88 | ||
| AK165976.1 | - | 7 - 8 | 1099 | 88 | ||
| AK167222.1 | - | 7 - 8 | 1113 | 88 | ||
| BC052648.1 | - | 7 - 8 | 1065 | 30030801 | 88 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trmt6
Genomic Map