Assay Details
| Gene Symbol: Lcmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025304.3 | NP_079580.2 | 4 - 5 | 566 | 146 | |
| GenBank mRNA | BC132507.1 | - | 4 - 5 | 472 | 40130784 | 146 |
| BC132509.1 | - | 4 - 5 | 472 | 40130786 | 146 | |
| BC145301.1 | - | 4 - 5 | 472 | 146 | ||
More Information
Set Membership: |
|

Lcmt1
Genomic Map