Assay Details
| Gene Symbol: Cox6a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009943.2 | NP_034073.2 | 1 - 2 | 149 | 70 | |
| GenBank mRNA | AK002506.1 | - | 1 - 2 | 100 | 70 | |
| BC028514.1 | - | 1 - 2 | 120 | 1447375 | 70 | |
| BU842968.1 | - | 1 - 2 | 149 | 70 | ||
| U08439.1 | - | 1 - 2 | 135 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cox6a2
Genomic Map