Assay Details
| Gene Symbol: P2rx3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145526.2 | NP_663501.2 | 8 - 9 | 1384 | 61 | |
| GenBank mRNA | AK140283.1 | - | 8 - 9 | 2652 | 61 | |
| AK141847.1 | - | 8 - 9 | 1049 | 61 | ||
| BC023089.1 | - | 8 - 9 | 1037 | 3984739 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

P2rx3
Genomic Map