Assay Details
| Gene Symbol: Stk32c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001162540.1 | NP_001156012.1 | 4 - 5 | 480 | 74 | |
| NM_021302.3 | NP_067277.2 | 4 - 5 | 780 | 74 | ||
| GenBank mRNA | AB041542.1 | - | 4 - 5 | 760 | 74 | |
| AK046439.1 | - | 4 - 5 | 758 | 74 | ||
| BC026457.1 | - | 4 - 5 | 765 | 4481218 | 74 | |
More Information
Updated Mapping Information
Set Membership: |
|

Stk32c
Genomic Map