Assay Details
| Gene Symbol: Cd19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009844.2 | NP_033974.2 | 6 - 7 | 1025 | 80 | |
| GenBank mRNA | AK089835.1 | - | 6 - 7 | 1024 | 80 | |
| AK156125.1 | - | 6 - 7 | 1025 | 80 | ||
| AK163608.1 | - | 4 - 5 | 1522 | 80 | ||
| AK172008.1 | - | 6 - 7 | 987 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd19
Genomic Map