Assay Details
| Gene Symbol: Cd19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009844.2 | NP_033974.2 | 9 - 10 | 1320 | 69 | |
| GenBank mRNA | AK156125.1 | - | 9 - 10 | 1320 | 69 | |
| AK163608.1 | - | 7 - 8 | 1817 | 69 | ||
| AK172008.1 | - | 9 - 10 | 1282 | 69 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cd19
Genomic Map