Assay Details
| Gene Symbol: Pot1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028370.1 | NP_082646.1 | 14 - 15 | 1810 | 148 | |
| GenBank mRNA | AK045286.1 | - | 14 - 15 | 1820 | 148 | |
| AK050727.1 | - | 14 - 15 | 1809 | 148 | ||
| AK165040.1 | - | 14 - 15 | 1787 | 148 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pot1b
Genomic Map