Assay Details
| Gene Symbol: Pot1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028370.1 | NP_082646.1 | 1 - 2 | 82 | 139 | |
| GenBank mRNA | AK013364.1 | - | 1 - 2 | 83 | 139 | |
| AK050727.1 | - | 1 - 2 | 81 | 139 | ||
| AK165040.1 | - | 1 - 2 | 59 | 139 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK165040.1, NM_028370.1 transcripts.
Set Membership: |
|

Pot1b
Genomic Map