Assay Details
| Gene Symbol: March2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252480.1 | NP_001239409.1 | 4 - 5 | 764 | 68 | |
| NM_145486.5 | NP_663461.2 | 4 - 5 | 764 | 68 | ||
| NR_045522.1 | 4 - 5 | 764 | 68 | |||
| GenBank mRNA | AK039778.1 | - | 4 - 5 | 757 | 68 | |
| AK053434.1 | - | 4 - 5 | 732 | 68 | ||
| AK079234.1 | - | 4 - 5 | 741 | 68 | ||
| AK081495.1 | - | 4 - 5 | 742 | 68 | ||
| AK181591.1 | - | 2 - 3 | 229 | 68 | ||
| AK189593.1 | - | 2 - 3 | 229 | 68 | ||
| BC002144.1 | - | 4 - 5 | 724 | 3484751 | 68 | |
| BC132541.1 | - | 3 - 4 | 640 | 40130818 | 68 | |
| BC138064.1 | - | 3 - 4 | 640 | 8861083 | 68 | |
More Information
Set Membership: |
|

March2
Genomic Map