Assay Details
| Gene Symbol: Wdr46 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020603.2 | NP_065628.1 | 10 - 11 | 1127 | 119 | |
| GenBank mRNA | AK145068.1 | - | 10 - 11 | 1182 | 119 | |
| BC013527.1 | - | 10 - 11 | 1170 | 119 | ||
| BC026431.1 | - | 7 - 8 | 751 | 119 | ||
| BC046977.1 | - | 10 - 11 | 1127 | 6315172 | 119 | |
More Information
Set Membership: |
|

Wdr46
Genomic Map